Italy
Order Entry
Dispersing aggregates for POLYTRON® system PT10-35 GT
Dispersing aggregates for POLYTRON® system PT10-35 GT
undefined
Dispersing aggregates for POLYTRON® system PT10-35 GT
Standard dispersing aggregate
  •  
    Speed
    Volume
    Pk
  • PT-DA 07/2EC-F101, Ø×H 7×101 mm
    PT-DA 07/2EC-F101, Ø×H 7×101 mm
    KINE11030298
    11030298
    8 m/s
    0,3 - 10 ml
    1
    1 * 1 pezzi
    1 * 1 pezzi - 2 010,00 €
    2 010,00 €
    1 * 1 pezzi
       2 010,00 €
    Specifications
    Environmentally Preferable
  • PT-DA 30/2EC-F250, Ø×H 30×250 mm
    PT-DA 30/2EC-F250, Ø×H 30×250 mm
    KINE11030305
    11030305
    30 m/s
    100 - 4000 ml
    1
    1 * 1 pezzi
    1 * 1 pezzi - 2 930,00 €
    2 930,00 €
    1 * 1 pezzi
       2 930,00 €
    Specifications
    Environmentally Preferable
  • PT-DA 36/4EC-F250, Ø×H 36×250 mm
    PT-DA 36/4EC-F250, Ø×H 36×250 mm
    KINE11030317
    11030317
    34 m/s
    200 - 4500 ml
    1
    1 * 1 pezzi
    1 * 1 pezzi - 3 890,00 €
    3 890,00 €
    1 * 1 pezzi
       3 890,00 €
    Specifications
    Environmentally Preferable
  • PT-DA 50/4EC-F250, Ø×H 50×250 mm
    PT-DA 50/4EC-F250, Ø×H 50×250 mm
    KINE11030335
    11030335
    44 m/s
    300 - 10000 ml
    1
    1 * 1 pezzi
    1 * 1 pezzi - 4 310,00 €
    4 310,00 €
    1 * 1 pezzi
       4 310,00 €
    Specifications
    Environmentally Preferable
  • PT-DA 60/2EC-F250, Ø×H 60×250 mm
    PT-DA 60/2EC-F250, Ø×H 60×250 mm
    KINE11030338
    11030338
    44 m/s
    300 - 30000 ml
    1
    1 * 1 pezzi
    1 * 1 pezzi - 4 290,00 €
    4 290,00 €
    1 * 1 pezzi
       4 290,00 €
    Specifications
    Environmentally Preferable
W-Design
  •  
    Speed
    Volume
    Pk
  • PT-DA 20/2WEC-F193, Ø×H 20×193 mm
    PT-DA 20/2WEC-F193, Ø×H 20×193 mm
    KINE11030304
    11030304
    22 m/s
    20 - 2000 ml
    1
    1 * 1 pezzi
    1 * 1 pezzi - 2 770,00 €
    2 770,00 €
    1 * 1 pezzi
       2 770,00 €
    Specifications
    Environmentally Preferable
  • PT-DA 36/2WEC-F250, Ø×H 36×250 mm
    PT-DA 36/2WEC-F250, Ø×H 36×250 mm
    KINE11030319
    11030319
    34 m/s
    200 - 4500 ml
    1
    1 * 1 pezzi
    1 * 1 pezzi - 3 890,00 €
    3 890,00 €
    1 * 1 pezzi
       3 890,00 €
    Specifications
    Environmentally Preferable
  • PT-DA 12/2WEC-F154, Ø×H 12×154 mm
    PT-DA 12/2WEC-F154, Ø×H 12×154 mm
    KINE11030301
    11030301
    14 m/s
    3 - 250 ml
    1
    1 * 1 pezzi
    1 * 1 pezzi - 2 500,00 €
    2 500,00 €
    1 * 1 pezzi
       2 500,00 €
    Specifications
    Environmentally Preferable
  • PT-DA 30/2WEC-F250, Ø×H 30×250 mm
    PT-DA 30/2WEC-F250, Ø×H 30×250 mm
    KINE11030309
    11030309
    30 m/s
    100 - 4000 ml
    1
    1 * 1 pezzi
    1 * 1 pezzi - 3 580,00 €
    3 580,00 €
    1 * 1 pezzi
       3 580,00 €
    Specifications
    Environmentally Preferable
Biotrona® B-Design high-turbulence mixer
  •  
    Speed
    Volume
    Pk
  • PT-DA 20/BEC-F193, Ø×H 20×193 mm
    PT-DA 20/BEC-F193, Ø×H 20×193 mm
    KINE11030296
    11030296
    21 m/s
    100 - 2000 ml
    1
    1 * 1 pezzi
    1 * 1 pezzi - 2 480,00 €
    2 480,00 €
    1 * 1 pezzi
       2 480,00 €