Consumables for Shimadzu autosamplers
Supplier: Trajan Scientific and Medical
Certificates
About this item
Autosampler syringes for AOC14, AOC17 and AOC20.
* Gas-tight.
Related Information
Autosampler syringes
Product Information
- Catalog #Supplier #VolumeTypeType of tipGaugeExt.ØAvailabilityPrice
- Removable needleRemovable needle549-04100004400,5 µl0.5BR-S-0.47Ccone260,47 mmLog in for availability1 * 1 pezzi196,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - Removable needleRemovable needle562-103800289710 µl10R-S-0.47Ccone260,47 mmLog in for availability1 * 1 pezzi131,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - Removable needleRemovable needle562-103900289810 µl10R-S-0.63Ccone230,63 mmLog in for availability1 * 1 pezzi131,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - Fixed needleSpecifications:
Accessories
Specifications
Environmentally Preferable - Removable needle *Removable needle *549-041300290210 µl10BR-S-GT-0.63Ccone230,63Log in for availability1 * 1 pezzi142,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - Removable needleRemovable needle549-04110004450,5 µl0.5BR-S-0.63Ccone230,63 mmLog in for availability1 * 1 pezzi196,00 €Specifications:
Accessories
Specifications
Environmentally Preferable
Spare needles for autosampler syringes
Product Information
- Catalog #Supplier #VolumeTypeType of tipGaugeExt.ØAvailabilityPrice
- Specifications:
Accessories
Specifications
Environmentally Preferable - Specifications:
Accessories
Specifications
Environmentally Preferable - Specifications:
Accessories
Specifications
Environmentally Preferable - Specifications:
Accessories
Specifications
Environmentally Preferable - Specifications:
Accessories
Specifications
Environmentally Preferable
Inlet liners
Product Information
- Catalog #Supplier #VolumeTypeType of tipGaugeExt.ØAvailabilityPrice
- Ferrule liner 17ASpecifications:
Accessories
Specifications
Environmentally Preferable - O-ring for Liner 2010Specifications:
Accessories
Specifications
Environmentally Preferable - For 2010 (SPL-2010 injector), 3 baffle for PTVFor 2010 (SPL-2010 injector), 3 baffle for PTVSGEA0920425010920425013,1 mmLog in for availability1 * 1 pezzi125,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 2010 (SPL-2010 injector), 3 baffle for PTVFor 2010 (SPL-2010 injector), 3 baffle for PTVSGEA092042509204253,1 mmLog in for availability1 * 5 pezzi493,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 2010 (SPL-2010 injector), tapered FocusLiner™, split/splitlessFor 2010 (SPL-2010 injector), tapered FocusLiner™, split/splitless558-0602092058255,0 mmLog in for availability1 * 25 pezzi1 550,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 2010 (SPL-2010 injector), FocusLiner™, split/splitlessFor 2010 (SPL-2010 injector), FocusLiner™, split/splitless558-0601092059255,0 mmLog in for availability1 * 25 pezzi1 100,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 2010 (SPL-2010 injector), FocusLiner™, split/splitlessFor 2010 (SPL-2010 injector), FocusLiner™, split/splitless558-04920920595,0 mmLog in for availability1 * 5 pezzi258,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2014, split/splitless with gooseneck & quartz woolFor 17A (SP-17 injector) & 2014, split/splitless with gooseneck & quartz woolSGEA09206101092061015,0 mmLog in for availability1 * 1 pezzi59,90 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2014, FocusLiner™, split/splitlessFor 17A (SP-17 injector) & 2014, FocusLiner™, split/splitlessSGEA09206201092062015,0 mmLog in for availability1 * 1 pezzi64,60 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 2010 (SPL-2010 injector), split/splitless with middle gooseneckFor 2010 (SPL-2010 injector), split/splitless with middle gooseneck558-04560920775,0 mmLog in for availability1 * 5 pezzi201,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 14, 15A & 16 (SPL-14 injector), split/splitless with 2 mm gooseneckFor 14, 15A & 16 (SPL-14 injector), split/splitless with 2 mm gooseneck558-04570920825,0 mmLog in for availability1 * 5 pezzi224,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 14, 15A & 16 (SPL-14 injector), split/splitless with single taperFor 14, 15A & 16 (SPL-14 injector), split/splitless with single taper558-049409208315,0 mmLog in for availability1 * 5 pezzi231,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 14, 15A & 16 (SPL-14 injector), splitless, direct wide bore for 9A & 16 A GC'sFor 14, 15A & 16 (SPL-14 injector), splitless, direct wide bore for 9A & 16 A GC's558-04950920845,0 mmLog in for availability1 * 5 pezzi313,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2014, split/splitless with middle gooseneckFor 17A (SP-17 injector) & 2014, split/splitless with middle gooseneck558-04960920855,0 mmLog in for availability1 * 5 pezzi213,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2010, splitless, straight throughFor 17A (SP-17 injector) & 2010, splitless, straight throughSGEA0920861010920861015,0 mmLog in for availability1 * 1 pezzi30,90 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2010, direct, for column 0.53 mm IDFor 17A (SP-17 injector) & 2010, direct, for column 0.53 mm IDSGEA09208701092087015,0 mmLog in for availability1 * 1 pezzi90,50 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2014, SPM linerFor 17A (SP-17 injector) & 2014, SPM liner558-04590920895,0 mmLog in for availability1 * 5 pezzi207,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 2010 (SPL-2010 injector), straight, non-sintered, with wool for LVIFor 2010 (SPL-2010 injector), straight, non-sintered, with wool for LVISGEA09229201092292013,1 mmLog in for availability1 * 1 pezzi85,60 €Specifications:
Accessories
Specifications
Environmentally Preferable - ConnecTite liner top holeSpecifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2014, SPM linerFor 17A (SP-17 injector) & 2014, SPM linerSGEA09208901092089015,0 mmLog in for availability1 * 1 pezzi51,80 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 2010 (SPL-2010 injector), straight, non-sintered, with wool for LVIFor 2010 (SPL-2010 injector), straight, non-sintered, with wool for LVISGEA0922920922923,1 mmLog in for availability1 * 5 pezzi339,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - ConnecTite liner standardSpecifications:
Accessories
Specifications
Environmentally Preferable - ConnecTite liner bottom holeSpecifications:
Accessories
Specifications
Environmentally Preferable - For 2010 (SPL-2010 injector), tapered FocusLiner™, split/splitlessFor 2010 (SPL-2010 injector), tapered FocusLiner™, split/splitlessSGEA09205801092058015,0 mmLog in for availability1 * 1 pezzi92,20 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 2010 (SPL-2010 injector), tapered FocusLiner™, split/splitlessFor 2010 (SPL-2010 injector), tapered FocusLiner™, split/splitless558-04500920585,0 mmLog in for availability1 * 5 pezzi365,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 2010 (SPL-2010 injector), FocusLiner™, split/splitlessFor 2010 (SPL-2010 injector), FocusLiner™, split/splitlessSGEA09205901092059015,0 mmLog in for availability1 * 1 pezzi64,60 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2014, split/splitless with gooseneck & quartz woolFor 17A (SP-17 injector) & 2014, split/splitless with gooseneck & quartz wool558-04510920615,0 mmLog in for availability1 * 5 pezzi234,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2014, FocusLiner™, split/splitlessFor 17A (SP-17 injector) & 2014, FocusLiner™, split/splitless558-0603092062255,0 mmLog in for availability1 * 25 pezzi1 100,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2014, FocusLiner™, split/splitlessFor 17A (SP-17 injector) & 2014, FocusLiner™, split/splitless562-10880920625,0 mmLog in for availability1 * 5 pezzi258,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 2010 (SPL-2010 injector), split , straigh throughFor 2010 (SPL-2010 injector), split , straigh throughSGEA09206401092064015,0 mmLog in for availability1 * 1 pezzi42,30 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 2010 (SPL-2010 injector), split , straigh throughFor 2010 (SPL-2010 injector), split , straigh through558-04530920645,0 mmLog in for availability1 * 5 pezzi162,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 14, 15A & 16 (SPL-14 injector), FocusLiner™, split/splitlessFor 14, 15A & 16 (SPL-14 injector), FocusLiner™, split/splitlessSGEA09206501092065015,0 mmLog in for availability1 * 1 pezzi63,20 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 14, 15A & 16 (SPL-14 injector), FocusLiner™, split/splitlessFor 14, 15A & 16 (SPL-14 injector), FocusLiner™, split/splitless558-04930920655,0 mmLog in for availability1 * 5 pezzi252,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 14, 15A & 16 (SPL-14 injector), tapered FocusLiner™, split/splitlessFor 14, 15A & 16 (SPL-14 injector), tapered FocusLiner™, split/splitless558-04540920665,0 mmLog in for availability1 * 5 pezzi334,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2014, tapered FocusLiner™, split/splitlessFor 17A (SP-17 injector) & 2014, tapered FocusLiner™, split/splitlessSGEA09206801092068015,0 mmLog in for availability1 * 1 pezzi92,20 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2014, tapered FocusLiner™, split/splitlessFor 17A (SP-17 injector) & 2014, tapered FocusLiner™, split/splitless558-0604092068255,0 mmLog in for availability1 * 25 pezzi1 550,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2014, tapered FocusLiner™, split/splitlessFor 17A (SP-17 injector) & 2014, tapered FocusLiner™, split/splitless558-03180920685,0 mmLog in for availability1 * 5 pezzi365,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 2014 and 2010 injectors, tapered FocusLiner™, split/splitlessFor 2014 and 2010 injectors, tapered FocusLiner™, split/splitlessSGEA09207101092071015,0 mmLog in for availability1 * 1 pezzi102,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 2014 and 2010 injectors, tapered FocusLiner™, split/splitlessFor 2014 and 2010 injectors, tapered FocusLiner™, split/splitless558-04550920715,0 mmLog in for availability1 * 5 pezzi405,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 14, 15A & 16 (SPL-14 injector), splitless, direct wide bore for 9A & 16 A GC'sFor 14, 15A & 16 (SPL-14 injector), splitless, direct wide bore for 9A & 16 A GC'sSGEA09208401092084015,0 mmLog in for availability1 * 1 pezzi79,20 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2014, split/splitless with middle gooseneckFor 17A (SP-17 injector) & 2014, split/splitless with middle gooseneckSGEA09208501092085015,0 mmLog in for availability1 * 1 pezzi53,30 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2014, split/splitless with middle gooseneckFor 17A (SP-17 injector) & 2014, split/splitless with middle gooseneck558-0609092085255,0 mmLog in for availability1 * 25 pezzi900,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2010, splitless, straight throughFor 17A (SP-17 injector) & 2010, splitless, straight through558-045809208615,0 mmLog in for availability1 * 5 pezzi118,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - For 17A (SP-17 injector) & 2010, direct, for column 0.53 mm IDFor 17A (SP-17 injector) & 2010, direct, for column 0.53 mm ID558-04970920875,0 mmLog in for availability1 * 5 pezzi355,00 €Specifications:
Accessories
Specifications
Environmentally Preferable
Ferrules for GC14A, GC17A, GC2010 and GC2014 detector/injectors (not for MS interfaces or QP2010 injector)
Product Information
- Catalog #Supplier #VolumeTypeType of tipGaugeExt.ØAvailabilityPrice
- 5 mm OD columns packedSpecifications:
Accessories
Specifications
Environmentally Preferable - 0,45 - 0,53 mm ID columnsSpecifications:
Accessories
Specifications
Environmentally Preferable - 0,10 - 0,32 mm ID columnsSpecifications:
Accessories
Specifications
Environmentally Preferable - Replacement ferrules, column ID 1/32"Specifications:
Accessories
Specifications
Environmentally Preferable - Replacement ferrules, column ID 0,53 mmReplacement ferrules, column ID 0,53 mm558-0429073229Log in for availability1 * 10 pezzi145,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - NutSpecifications:
Accessories
Specifications
Environmentally Preferable - SilTite™ kit (2 nuts, 10 ferrules) for 0,10 - 0,25 mm ID columnsSilTite™ kit (2 nuts, 10 ferrules) for 0,10 - 0,25 mm ID columns558-0486073227Log in for availability1 * 10 pezzi145,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - SilTite™ kit (2 nuts, 10 ferrules), column ID 0,10 - 0,25 mmSilTite™ kit (2 nuts, 10 ferrules), column ID 0,10 - 0,25 mm558-0265073350Log in for availability1 * 1 pezzi163,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - SilTite™ kit (2 nuts, 10 ferrules), column ID 0,32 mmSilTite™ kit (2 nuts, 10 ferrules), column ID 0,32 mm558-0266073351Log in for availability1 * 1 pezzi163,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - SilTite™ kit (2 nuts, 10 ferrules), column ID 0,45 - 0,53 mmSilTite™ kit (2 nuts, 10 ferrules), column ID 0,45 - 0,53 mm558-0267073352Log in for availability1 * 1 pezzi163,00 €Specifications:
Accessories
Specifications
Environmentally Preferable
Ferrules for QP5000/5050 standard MS interface
Product Information
- Catalog #Supplier #VolumeTypeType of tipGaugeExt.ØAvailabilityPrice
- SilTite™ kit (2 nuts, 10 ferrules) for 0,32 mm ID columnsSilTite™ kit (2 nuts, 10 ferrules) for 0,32 mm ID columns558-0424073205Log in for availability1 * 1 pezzi163,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - SilTite™ kit (2 nuts, 10 ferrules) for 0,10 - 0,25 mm ID columnsSilTite™ kit (2 nuts, 10 ferrules) for 0,10 - 0,25 mm ID columns558-0423073204Log in for availability1 * 1 pezzi163,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - Nuts for QP5000 Direct MS interfaceSpecifications:
Accessories
Specifications
Environmentally Preferable - QP5000-II & QP5050 0,32 mm ID columnsSpecifications:
Accessories
Specifications
Environmentally Preferable - QP5000-II & QP5050 0,10 - 0,25 mm ID columnsQP5000-II & QP5050 0,10 - 0,25 mm ID columns558-04190726561Log in for availability1 * 10 pezzi149,00 €Specifications:
Accessories
Specifications
Environmentally Preferable - QP5000-I 0,32 mm ID columnsSpecifications:
Accessories
Specifications
Environmentally Preferable - QP5000-I 0,10 - 0,25 mm ID columnsSpecifications:
Accessories
Specifications
Environmentally Preferable
Spare needles for autosampler syringes
Product Information
- Catalog #SeqDescriptionGaugeLengthExt.ØØ int.Type of tipTypeAvailabilityPrice
- 33Spare needle and plunger kit23-2642 mm0,63/0,47 mm0,10 mmConeNP0.5B-AG-0.63/0.47Log in for availability
Ferrules for GC14A, GC17A, GC2010 and GC2014 detector/injectors (not for MS interfaces or QP2010 injector)
Product Information
- Catalog #SeqDescriptionTypeGaugeØ int.MaterialPkPriceAvailabilityPrice
- 45For injectors & detectors at atm. pressure e.g FID, column ID 0, 45 - 0,53 mm (1/8" nut)OGF-PE-010J0,8 mmGraphite1023,00Log in for availability
Ferrules for QP5000/5050 standard MS interface
Product Information
- Catalog #SeqDescriptionMaterialPkPriceAvailabilityPrice
Ferrules for QP5000/5050 wide bore MS interface, QP2010 injector and QP2010 s
Product Information
- Catalog #SeqDescriptionØ int.PkPriceAvailabilityPrice
- 120For GC-MS interface connection, column ID 0,1 - 0,25 mm, material: SilTite™ starter kit0,4 mm153,00Log in for availability