Stampa…
Airline filtration unit, AFU 300
Fornitore: Scott Safety
SCTS2004876EA 585 EUR
SCTS2004876
Airline filtration unit, AFU 300
AFU series of compressed airline filtration units offer an economical means of ensuring compressed air conforming to breathing quality requirements.
  • Effective two stage filtration of particulates including dust and oil droplets
  • Charcoal element for removal of organic vapours
  • Pop-up indicator to warn of prefilter clogging
  • Autodrain to prevent build-up of oil
  • Easy filter element replacement without use of tools
These units have the capacity to supply air flows to the latest recommendations for breathing apparatus and will assist in ensuring air quality. These are not suitable for carbon monoxide or carbon dioxide in the air supply.
Certificazioni: Meets EN 12021 and BS4275:1997 requirements
Informazioni per l'ordine: Includes lightweight and robust carry frame.