HCV NS3 genotype-5
: Biorbyt
- Storage conditions:Five years frozen. One month in solution at room temperature.
- Protein/peptide name:HCV NS3 Genotype-5
- Purity:HCV NS3 Genotype-5 protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV NS3 Genotype-5 protein was purified by proprietary chromatographic technique.
- Formulation:1.5M urea, 25mM Tris-HCl pH-8, 0.2% Triton-X 50% Glycerol.
- Tested applications:SDS-PAGE
- Pk
- : ORB81587-100: BIRBORB81587-100
- : ORB81587-500: BIRBORB81587-500