Italy
Order Entry
Hygiene masks, PP
  114-9101
Hygiene masks, PP
  114-9101
 : HOIT114531
  1 * 50 pezzi
  • Type:
    With earloops
  • Colour:
    Green
  • Seq:
    02
  • Pk:
    50
Hygiene masks made from 3-ply construction of PP and Meltblown PP.
  • Easy breathing, non-irritating
  • Excellent bacterial and particle filtration
  • Full length aluminum nose piece for a comfortable fit
  • For industrial and clean room use only; do not use for protection against COVID-19
  • This product is not a medical device for medical or clinical use within the meaning of Regulation (EU) 2017/745 / Directive 93/42/EEC
  • This product is not a personal protective equipment (PPE) within the meaning of Regulation (EU) 2016/425