Stampa…
Isolation gowns
Fornitore: Hubei Orient
HOIT110412EA 80.5 EUR
HOIT110412
Isolation gowns
Indumenti usa e getta
Full length gown made of soft PP fabric.
  • Elastic cuffs
  • Extra long ties sewn at waist and neckline
  • Medical repellence grade (anti-alcohol, anti-blood, anti-oil), anti-static
  • No dust and particle release