Italy
Order Entry
Cell culture flasks, Corning®
 :  Corning
undefined
Cell culture flasks, Corning®
 :  Corning
  • Catalog No:
  • 734-1718
  • 734-4139
  • 734-1214
  • 734-1726
  • 734-0092
  • 734-0094
  • 734-2272
  • 734-0091
  • 734-2274
  • 734-1712
  • 734-1733
  • 734-2275
  • 734-1706
  • 734-1725
  • 734-0093
  • 734-1724
  • 734-1723
  • 734-1722
  • 734-0090
  • 734-1540
  • 734-1700
  • 734-1717
  • 734-2708
  • 734-2707
  • 734-2706
  • 734-2705
  • 734-1719
  • 734-1715
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
25 cm² Growth area flasks
  •  
    Capacity
    Growth area
    Closure type
    Neck
    Recommended working volume
  • Non-treated, rectangular flasks
    Non-treated, rectangular flasks
    734-2272
    431463
    10 ml
    25 cm²
    Filter cap
    Canted
    5-7,5 ml
    1 * 200 pezzi
    1 * 200 pezzi - 283,77 €
    283,77 €
    1 * 200 pezzi
       283,77 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • TC-treated, rectangular flasks
    TC-treated, rectangular flasks
    734-1712
    430639
    10 ml
    25 cm²
    Filter cap
    Canted
    5-7,5 ml
    1 * 200 pezzi
    1 * 200 pezzi - 341,87 €
    341,87 €
    1 * 200 pezzi
       341,87 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • TC-treated, rectangular flasks
    TC-treated, rectangular flasks
    734-1706
    430372
    10 ml
    25 cm²
    Cap, phenolic
    Canted
    5-7,5 ml
    1 * 500 pezzi
    1 * 500 pezzi - 688,12 €
    688,12 €
    1 * 500 pezzi
       688,12 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • CellBIND® surface treatment, rectangular flasks
    CellBIND® surface treatment, rectangular flasks
    734-0090
    3289
    10 ml
    25 cm²
    Filter cap
    Canted
    5-7,5 ml
    1 * 200 pezzi
    1 * 200 pezzi - 458,00 €
    458,00 €
    1 * 200 pezzi
       458,00 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • TC-treated, rectangular flasks
    TC-treated, rectangular flasks
    734-1700
    430168
    10 ml
    25 cm²
    Plug seal screw cap
    Canted
    5-7,5 ml
    1 * 500 pezzi
    1 * 500 pezzi - 688,12 €
    688,12 €
    1 * 500 pezzi
       688,12 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
75 cm² Growth area flasks
  •  
    Capacity
    Growth area
    Closure type
    Neck
    Recommended working volume
  • Ultra-low attachment treated, U-shaped flasks
    Ultra-low attachment treated, U-shaped flasks
    734-4139
    3814
    60 ml
    75 cm²
    Filter cap
    Canted
    15-22,5 ml
    1 * 24 pezzi
    1 * 24 pezzi - 471,54 €
    471,54 €
    1 * 24 pezzi
       471,54 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • CellBIND® surface treatment , U-shaped flasks
    CellBIND® surface treatment , U-shaped flasks
    734-0091
    3290
    60 ml
    75 cm²
    Filter cap
    Canted
    15-22,5 ml
    1 * 100 pezzi
    1 * 100 pezzi - 444,00 €
    444,00 €
    1 * 100 pezzi
       444,00 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • TC-treated, U-shape flasks
    TC-treated, U-shape flasks
    734-2705
    430641U
    45 ml
    75 cm²
    Vented cap
    Canted
    15-22,5 ml
    1 * 100 pezzi
    1 * 100 pezzi - 330,51 €
    330,51 €
    1 * 100 pezzi
       330,51 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • TC-treated, rectangular flasks
    TC-treated, rectangular flasks
    734-1715
    430720
    45 ml
    75 cm²
    Plug seal screw cap
    Canted
    15-22,5 ml
    1 * 100 pezzi
    1 * 100 pezzi
    Specifications
    Environmentally Preferable
    Environmental certification
  • TC-treated, U-shaped flasks
    TC-treated, U-shaped flasks
    734-2706
    430720U
    45 ml
    75 cm²
    Plug seal screw cap
    Canted
    15-22,5 ml
    1 * 100 pezzi
    1 * 100 pezzi - 286,35 €
    286,35 €
    1 * 100 pezzi
       286,35 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • TC-treated, U-shaped flasks
    TC-treated, U-shaped flasks
    734-2707
    430725U
    45 ml
    75 cm²
    Cap, phenolic
    Canted
    15-22,5 ml
    1 * 100 pezzi
    1 * 100 pezzi - 287,30 €
    287,30 €
    1 * 100 pezzi
       287,30 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • Non-treated, U-shaped flasks
    Non-treated, U-shaped flasks
    734-2708
    431464U
    45 ml
    75 cm²
    Vented cap
    Canted
    15-22,5 ml
    1 * 100 pezzi
    1 * 100 pezzi - 270,39 €
    270,39 €
    1 * 100 pezzi
       270,39 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
150 cm² Growth area flasks
  •  
    Capacity
    Growth area
    Closure type
    Neck
    Recommended working volume
  • TC-treated, rectangular flasks
    TC-treated, rectangular flasks
    734-1718
    430824
    210 ml
    150 cm²
    Cap, phenolic
    Canted
    30-45 ml
    1 * 50 pezzi
    1 * 50 pezzi - 248,24 €
    248,24 €
    1 * 50 pezzi
       248,24 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • CellBIND® surface treatment, rectangular flasks
    CellBIND® surface treatment, rectangular flasks
    734-0092
    3291
    210 ml
    150 cm²
    Filter cap
    Canted
    30-45 ml
    1 * 50 pezzi
    1 * 50 pezzi - 370,00 €
    370,00 €
    1 * 50 pezzi
       370,00 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • Non-treated, rectangular flasks
    Non-treated, rectangular flasks
    734-2274
    431465
    210 ml
    150 cm²
    Filter cap
    Canted
    30-45 ml
    1 * 50 pezzi
    1 * 50 pezzi - 230,58 €
    230,58 €
    1 * 50 pezzi
       230,58 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • TC-treated, rectangular flasks
    TC-treated, rectangular flasks
    734-1717
    430823
    210 ml
    150 cm²
    Plug seal screw cap
    Canted
    30-45 ml
    1 * 50 pezzi
    1 * 50 pezzi - 248,24 €
    248,24 €
    1 * 50 pezzi
       248,24 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • TC-treated, rectangular flasks
    TC-treated, rectangular flasks
    734-1719
    430825
    210 ml
    150 cm²
    Filter cap
    Canted
    30-45 ml
    1 * 50 pezzi
    1 * 50 pezzi - 276,42 €
    276,42 €
    1 * 50 pezzi
       276,42 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
162 cm² Growth area flasks
  •  
    Capacity
    Growth area
    Closure type
    Neck
    Recommended working volume
  • TC-treated, traditional flasks
    TC-treated, traditional flasks
    734-1540
    3150
    175 ml
    162 cm²
    Cap, phenolic
    Straight
    32-48 ml
    1 * 25 pezzi
    1 * 25 pezzi
    Specifications
    Environmentally Preferable
    Environmental certification
175 cm² Growth area flasks
  •  
    Capacity
    Growth area
    Closure type
    Neck
    Recommended working volume
  • CellBIND® surface treatment, rectangular flasks, with barcode
    CellBIND® surface treatment, rectangular flasks, with barcode
    734-1214
    431328
    250 ml
    175 cm²
    Filter cap
    Angled
    35-52,5 ml
    1 * 84 pezzi
    1 * 84 pezzi - 1 300,00 €
    1 300,00 €
    1 * 84 pezzi
       1 300,00 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • TC-treated, rectangular flasks
    TC-treated, rectangular flasks
    734-1726
    431085
    250 ml
    175 cm²
    Cap, phenolic
    Angled
    35-52,5 ml
    1 * 50 pezzi
    1 * 50 pezzi - 349,35 €
    349,35 €
    1 * 50 pezzi
       349,35 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • Non-treated, rectangular flasks
    Non-treated, rectangular flasks
    734-2275
    431466
    250 ml
    175 cm²
    Filter cap
    Angled
    35-52,5 ml
    1 * 50 pezzi
    1 * 50 pezzi - 326,39 €
    326,39 €
    1 * 50 pezzi
       326,39 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • TC-treated, rectangular flasks, with barcode
    TC-treated, rectangular flasks, with barcode
    734-1733
    431306
    250 ml
    175 cm²
    Filter cap
    Angled
    35-52,5 ml
    1 * 84 pezzi
    1 * 84 pezzi - 970,59 €
    970,59 €
    1 * 84 pezzi
       970,59 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • TC-treated, rectangular flasks
    TC-treated, rectangular flasks
    734-1722
    431079
    250 ml
    175 cm²
    Plug seal screw cap
    Angled
    35-52,5 ml
    1 * 50 pezzi
    1 * 50 pezzi - 349,35 €
    349,35 €
    1 * 50 pezzi
       349,35 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • TC-treated, rectangular flasks
    TC-treated, rectangular flasks
    734-1723
    431080
    250 ml
    175 cm²
    Filter cap
    Angled
    35-52,5 ml
    1 * 50 pezzi
    1 * 50 pezzi - 391,35 €
    391,35 €
    1 * 50 pezzi
       391,35 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • CellBIND® surface treatment, rectangular flasks
    CellBIND® surface treatment, rectangular flasks
    734-0093
    3292
    250 ml
    175 cm²
    Filter cap
    Angled
    35-52,5 ml
    1 * 50 pezzi
    1 * 50 pezzi - 524,00 €
    524,00 €
    1 * 50 pezzi
       524,00 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
225 cm² Growth area flasks
  •  
    Capacity
    Growth area
    Closure type
    Neck
    Recommended working volume
  • CellBIND® surface treatment, rectangular flasks
    CellBIND® surface treatment, rectangular flasks
    734-0094
    3293
    370 ml
    225 cm²
    Filter cap
    Angled
    45-67,5 ml
    1 * 25 pezzi
    1 * 25 pezzi - 370,00 €
    370,00 €
    1 * 25 pezzi
       370,00 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • TC-treated, rectangular flasks
    TC-treated, rectangular flasks
    734-1725
    431082
    370 ml
    225 cm²
    Filter cap
    Angled
    45-67,5 ml
    1 * 25 pezzi
    1 * 25 pezzi - 275,54 €
    275,54 €
    1 * 25 pezzi
       275,54 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact
  • TC-treated, rectangular flasks
    TC-treated, rectangular flasks
    734-1724
    431081
    370 ml
    225 cm²
    Plug seal screw cap
    Angled
    45-67,5 ml
    1 * 25 pezzi
    1 * 25 pezzi - 244,30 €
    244,30 €
    1 * 25 pezzi
       244,30 €
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
      Low Manufacturing Impact