Italy
Order Entry
Glass ceramic plates for wet digester system B-440
Glass ceramic plates for wet digester system B-440
Supplier:  BUCHI
Certificates
Glass ceramic plates for wet digester system B-440
Supplier:  BUCHI
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Glass ceramic plate, plain
    Glass ceramic plate, plain
    458-2214
    026982
    1
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi
    Specifications:
    Specifications
    Environmentally Preferable
  • Glass ceramic plate, 4 holes with Ø 48 mm
    Glass ceramic plate, 4 holes with Ø 48 mm
    455-0134
    036676
    1
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi
    Specifications:
    Specifications
    Environmentally Preferable
  • Glass ceramic plate, 4 holes with Ø 56 mm
    Glass ceramic plate, 4 holes with Ø 56 mm
    458-2213
    026663
    1
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi
    Specifications:
    Specifications
    Environmentally Preferable
  • Glass ceramic plate, 6 holes with Ø 39 mm
    Glass ceramic plate, 6 holes with Ø 39 mm
    458-2211
    026661
    1
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi
    Specifications:
    Specifications
    Environmentally Preferable