Italy
Order Entry
Anti-Human Nuclear Antigen Mouse Monoclonal Antibody [clone: 235-1]
Anti-Human Nuclear Antigen Mouse Monoclonal Antibody [clone: 235-1]
  BNUM0099-50
 :  Biotium
Anti-Human Nuclear Antigen Mouse Monoclonal Antibody [clone: 235-1]
  BNUM0099-50
 :  Biotium
 
417,00 €
   
  1 * 50 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    Human Nuclear Antigen
  • Clonality:
    Monoclonal
  • Clone:
    Clone 235-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Primates,
    Human
  • Antigen synonyms:
    HuNu|HNA
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

 

 

This MAb is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. MAb 235-1 recognizes an antigen associated with the nuclei in human cells. It can be used to stain the nuclei in cell or tissue preparations and can be used as a nuclear marker in subcellular fractions. It produces a speckled pattern in normal and malignant cells and may be used to stain the nuclei of cells in fixed or frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations.

  • Purified, BSA-free

Type: Primary
Antigen: Human Nuclear Antigen
Clonality: Monoclonal
Clone: 235-1
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Non-human primates