Stampa…
Cleaning trolley Clino®, CR4 EM-CR
Fornitore: Pfennig
PPSA3540600EA 1441.25 EUR
PPSA3540600
Cleaning trolley Clino®, CR4 EM-CR
The Clino® CR4 EM-CR trolley system made of stainless steel suitable for cleanrooms and for high sensitive areas that do not require sterility.
  • Individually configurable through Clino modular design system
  • EasyMop CR with ergonomically optimised system box
  • Highest efficiency and process reliability in single step technique
  • EasyMop system boxes available in red, green, blue, yeollow, grey and athracite
  • Includes contact-free dropping device MopDrop
  • Cloth impregnation for surface cleaning in PlusBox
  • Basic module mini with high end finish and castors (100 mm)
A small cleaning system with pre-preparation system EasyMop for technical cleanrooms suitable for ISO classes 7 and 8 or GMP areas C/D and it allows a validatable and reproducible results.
Informazioni per l'ordine: Supplied with 1×tray for Clino Systembox, 2×system boxes EasyMop CR, 2×PlusBoxes, disposal unit (1×35 L) and plastic container (5 L).