Stampa…
Anti-Chondroitin-6-Sulphate Mouse Monoclonal Antibody [clone: MK302]
Fornitore: MP Biomedicals
ICNA08636521EA 724 EUR
ICNA08636521
Anti-Chondroitin-6-Sulphate Mouse Monoclonal Antibody [clone: MK302]
Anticorpi
This mouse chondroitin 6 sulphate monoclonal antibody used in EIA, RIA and immunoblotting.
  • Dilution range: 1:100 to 1:5000 for ELISA
  • Dilution range: 1:100 to 1:200 for RIA
Type: Primary
Antigen: Chondroitin-6-Sulfate
Clonality: Monoclonal
Clone: MK302
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Dog, Human, Mouse, Rabbit, Rat, Sheep