Stampa…
Anti-Actin Mouse Monoclonal Antibody [clone: B4]
Fornitore: MP Biomedicals
ICNA08691331EA 448 EUR
ICNA08691331
Anti-Actin Mouse Monoclonal Antibody [clone: B4]
Anticorpi
  • Dilution range: 1:200 to 1:500
  • Tissue source: skeletal muscle
  • Incubation time: 2 hours
  • Temperature: room temperature
Type: Primary
Antigen: Actin
Clonality: Monoclonal
Clone: B4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human