Cleaning trolley Clino®, STEC7, with multiBox and plusBox
Supplier: Pfennig
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Clino®
PPSA2088640EA 65.32 GBP
PPSA2088640 PPSA2088660 PPSA3077670 PPSA2088622 PPSA2088611 PPSA2088677 PPSA2088666 PPSA2088644 PPSA2088633 PPSA2088610 PPSA2088620
Cleaning trolley Clino®, STEC7, with multiBox and plusBox
Trolleys
The STEC ergonomically optimised system trolley is exclusively developed and used for controlled areas. It is made of stainless steel, has enclosed appearance and open function with Clino® MultiBox and PlusBox.
  • Alternating between one and two-stage process is possible
  • Floor cleaning is simple with flat wringer Ringo and mop-reuse-technique
  • MultiBox with enormous storage space, extendable and lockable
  • PluBox system for the easy impregnation of mop covers and cloth (suitable for 6 l boxes)
  • Disposal unit at rest can be folded in, every time exchangeable and expandable
  • Double ball-bearinged guide wheels (125 mm)
MultiBox with its enclosed appearance and open function are used for the simple pre-preparation of mop heads and cloths. Both MultiBox and PlusBox are available in different colours.
Delivery information: Supplied with buckets (25 l and 15 l), bottle holder ClinoBar, 3×MultiBoxes, 4×PlusBoxes and disposal unit (2×70 l). PluBox includes a sieve and cover for higher safety in manual cleaning and a straight preparation of all textiles.
Order Now
Learn more