Visors, uvex pheos visor
Supplier: Uvex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
uvex pheos
111-1938EA 85.5 GBP
111-1938 111-1937 111-1939 111-1940 111-1936 111-1935
Visors, uvex pheos visor
Face Shields
Polycarbonate visor with uvex supravision excellence coating technology: Permanently anti-fog on the inside and extremely scratch resistant on the outside. Ideal for workplaces where you frequently move between indoor and outdoor environments, from light to dark.
  • Available with two alternative mountings: Mechanical side piece or automatic magnetic lock
  • Compatible with the uvex pheos E-S-WR and uvex pheos B-S-WR safety helmets, as well as with the uvex pheos alpine safety helmet for working at heights
  • Can be combined with uvex K2H and uvex K2H magnet earmuffs
Certified for areas of application 3 (liquids) and 9 (hot solids and molten metal)
pheos SLB 1: Certified in accordance with GS-ET 29 as Class 1 visor for protection from electrical arcs (4 kA)
pheos SLB 2: Certified in accordance with GS-ET 29 as Class 2 visor for protection from electrical arcs (7 kA), includes chin protector
Ordering information: Helmet not included. A visor adapter 9790.067 is required for combination with the uvex pheos alpine safety helmet.
Order Now
Learn more