Stampa…
Boats, tantalum
Fornitore: Thermo Fisher Scientific
41217.KUEA 141.44 EUR
41217.KU
Boats, tantalum
Navicelle Navicelle per combustione
Combustion boats, Tantalum.
For disc aperture cleaning, vacuum evaporation