Myricetin 95%
ACRO151590250
:
- Pack type:Glass bottle
- Pk:25 mg
- Seq:2315
- Appearance:Brown-yellow to green-brown Crystalline powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥94 %
- HPLC:≥98.5 %
:529-44-2
:
:C₁₅H₁₀O₈
:MFCD00006827
:318.24 g/mol
:>300 °C
:Ambient