Cold resistant gloves, uvex unilite thermo plus
Supplier: UVEX
Certificates
About this item
These winter safety gloves feature a dual-layer construction: The inner glove is made of acrylic and a new wool mix, the outer glove is made of polyamide/elastane. The gloves have a flexible polymer coating on palm, fingers and partially at the back of the hand.
- Extremely robust coating is flexible even at low temperatures
- Double layer construction ensures effective insulation
- Excellent dexterity, high flexibility
- Mechanical strength
Ideal for forklift truck drivers and for use in refrigerated warehouses or when working in cold conditions. For use in dry and slightly moist areas.
Certifications: EN 388 (3131X), EN 511 (010)
Specifications
- Catalog No:
- 112-4527
- 112-4526
- 112-4529
- 112-4528
- Model:
- plus
- plus
- plus
- plus
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Finish:
- Polymer coating on palm, fingers and back of the hand
- Polymer coating on palm, fingers and back of the hand
- Polymer coating on palm, fingers and back of the hand
- Polymer coating on palm, fingers and back of the hand
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388 (3131X), EN 511 (010)
- EN 388 (3131X), EN 511 (010)
- EN 388 (3131X), EN 511 (010)
- EN 388 (3131X), EN 511 (010)
Specifications
Product Information
- Catalog #Supplier #Glove sizeColourPkAvailabilityPrice
- Supplier Article Number: 6059208Local Article Number: UVEX6059208Specifications:
More
Specifications
ModelplusAmbidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableFinishPolymer coating on palm, fingers and back of the handCuff styleKnit wristEN normEN 388 (3131X), EN 511 (010) - Supplier Article Number: 6059209Local Article Number: UVEX6059209Specifications:
More
Specifications
ModelplusAmbidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableFinishPolymer coating on palm, fingers and back of the handCuff styleKnit wristEN normEN 388 (3131X), EN 511 (010) - Supplier Article Number: 6059210Local Article Number: UVEX6059210Specifications:
More
Specifications
ModelplusAmbidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableFinishPolymer coating on palm, fingers and back of the handCuff styleKnit wristEN normEN 388 (3131X), EN 511 (010) - Supplier Article Number: 6059211Local Article Number: UVEX6059211Specifications:
More
Specifications
ModelplusAmbidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableFinishPolymer coating on palm, fingers and back of the handCuff styleKnit wristEN normEN 388 (3131X), EN 511 (010)