Anti-Plzf Antibody
Catalog # BSBTPB9969
Supplier: Boster Bio
- Antibody type:Primary
- Antigen name:zinc finger and BTB domain containing 16
- Antigen symbol:ZBTB16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7704
- Antigen synonyms:PLZF|ZNF145
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Zinc finger and BTB domain-containing protein 16(ZBTB16) detection. Tested with WB in Human;Rat.
Type: Primary
Antigen: ZBTB16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat