Carrier proteins, Imject™ Blue Carrier™
Supplier: Thermo Fisher Scientific
Certificates
About this item
Imject™ Blue Carrier™ protein is a highly soluble, mollusc-derived hemocyanin that enable simple preparation of effective immunogens with amine- or carboxyl-peptide antigens.
- High-yield conjugation with each molecule of CCH contains hundreds of primary amines for coupling haptens via EDC or NHS ester crosslinkers
- Validated quality for the immunogenic carrier protein to ensure uniform chemical structure and solubility
- Superior solubility and highly immunogenic
Product Information
- Pk
- Imject™ Blue Carrier™ proteinImject™ Blue Carrier™ proteinPIER7713077130100 mg1 * 100 mg269,00 €
- Imject™ Blue Carrier™ protein
This product has been blocked by your organization. Please contact your purchasing department for more information