Stampa…
Foetal bovine serum, insect cell screened, US origin, HyClone™
Fornitore: HyClone products (Cytiva)
HYCLSH30070.03IEA 935 EUR
HYCLSH30070.03I
Foetal bovine serum, insect cell screened, US origin, HyClone™
HyClone™ foetal bovine serum, insect cell screened, is FBS that has been filtered through serial 40 nm pore-size rated filters and identified as batches that offer exceptional performance with baculovirus expression vector systems (BEVS).