Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Italy
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
GSK-3beta inhibitor VIII
GSK-3β inhibitor VIII
GSK-3β inhibitor VIII
GSK-3β inhibitor VIII
J64554.MA
:
Thermo Fisher Scientific
:
GSK-3β inhibitor VIII
J64554.MA
:
Thermo Fisher Scientific
:
: J64554.MA
: ALFAJ64554.MA
150,00 €
1 * 10 mg
-
+
Pk:
10 mg
Seq:
2275
:
:
308.31 g/mol
:
GHS05
:
GHS07
:
Refrigerator
:
487021-52-3
:
C₁₂H₁₂N₄O₄S
:
MFCD08277040