Italy
Order Entry
Phosphate-buffered saline (PBS, 5X), with EDTA
Phosphate-buffered saline (PBS, 5X), with EDTA
  • Physical form:
    Liquid
  •  
    Concentration
    Pk
  • Phosphate-buffered saline (PBS) with EDTA
    Phosphate-buffered saline (PBS) with EDTA
    J60893.K2
    J60893.K2
    5X
    1 L
    1 * 1 L
    1 * 1 L - 68,60 €
    68,60 €
    1 * 1 L
       68,60 €
  • Phosphate-buffered saline (PBS) with EDTA
    Phosphate-buffered saline (PBS) with EDTA
    J60893.K3
    J60893.K3
    5X
    2 L
    1 * 2 L
    1 * 2 L - 99,30 €
    99,30 €
    1 * 2 L
       99,30 €