Stampa…
pH/mV/°C meters, bench, Lab 855 / 865
Fornitore: SI Analytics
663-0326EA 1490 EUR
663-0326 663-0325 663-0313 663-0312
pH/mV/°C meters, bench, Lab 855 / 865
Misuratori elettrochimici pHmetri
The Lab 855 is ideal for measurements in laboratories in the chemical and pharmaceutical industries as well as in medical labs. The Lab 865 is perfect for pH measurements in quality assurance labs requiring the documentation for GLP. Built on the Lab 855 platform, the Lab 865 offers additional functionalities. Common features include:
  • Active AutoRead function with independent detection of stable measuring values for reproducible measuring results
  • Intuitive operation with clearly arranged keyboard
  • Simple calbration with adjustable calibration timer
Additional features of the Lab 865:
- CMC function (measuring range monitoring) for monitoring the congruency of measuring and calibration range for pH
- Data transfer via USB interface
- Manual or timer controlled data logging of measured values
- Protocols include date, time, and ID and sensors serial numbers for GLP compliance
- Data is transferred in *.csv format
- Excel Add-in included for the formatted output of all data and calibration protocols
Informazioni per l'ordine: Lab 855 (meter only without electrode) is supplied with power supply, batteries, stand and operating manual.
Lab 855 Set additionally includes pH combination electrode BlueLine 14 pH, calibration solutions and electrolyte solution.
Lab 865 (meter only without electrode) is supplied with power supply, batteries, stand, USB cable, software MultiLab Importer and operating manual.
Lab 865 Set additionally includes pH combination electrode BlueLine 14 pH and calibration solutions.