Stampa…
Evaluation software for all Ebro data loggers
Fornitore: Ebro
EBRO1340-2354EA 0 EUR
EBRO1340-2354
Evaluation software for all Ebro data loggers
Winlog.light is the standard software for programming and evaluating all Ebro data loggers.
  • Graphical and numeric display of the data progression
  • Different report types can be selected
  • Statistics for measured data (incl.MIN/MAX, mean value, standard deviation, etc.)
  • Data export to Microsoft® Excel
Required operating system: Microsoft® Windows® XP, Windows® 2000, Windows Vista™ or Windows™ 7