Specifications
- Antibody type:Primary
- Antigen name:PDZ Domain Containing 2
- Antigen symbol:PDZD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Gene ID:23037
- Antigen synonyms:PAPIN|PDZK3|PIN1|PDZ domain containing 3|PDZ domain-containing protein 3|PDZ domain-containing protein 2|AIPC|activated in prostate cancer protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Proteins containing PDZ domains have been shown frequently to bind the C-termini of transmembrane receptors or ion channels. They have also been shown to bind to other PDZ domain proteins and could possibly be involved in intracellular signalling. PDZK3 contains six PDZ domains and shares sequence similarity with pro-interleukin-16 (pro-IL-16). Like pro-IL-16, the encoded protein localizes to the endoplasmic reticulum and is thought to be cleaved by a caspase to produce a secreted peptide containing two PDZ domains. In addition, the PDZK3 gene is upregulated in primary prostate tumors and may be involved in the early stages of prostate tumorigenesis.
IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: PDZD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat