Stampa…
CO₂ incubators, BBK 6220
Fornitore: Thermo Scientific
390-0171EA 0 EUR
HERA50035416
CO₂ incubators, BBK 6220
The BBK 6220 with built-in refrigeration is a general purpose CO₂ incubator suitable for all applications which completes the 6000 range. Additional refrigeration function offers incubation at low temperatures. This feature is ideal in Marine biology and fish farming. The combination of refrigeration system and air jacket heating system provides optimum temperature and humidity.
  • Refrigeration unit with connection to a compact table top refrigeration thermostat
  • Microprocessor control
  • Automatic start-up and automatic auto-zero adjustment of CO₂ measuring cell
  • Independent electronic overtemperature protection with separate sensor
  • Heated external door and glass internal door
  • One-piece metal shelves
The BBK 6220 with built-in refrigeration is a general purpose CO² incubator suitable for all applications which completes the 6000 range. Additional refrigeration function offers incubation at low temperatures. This feature is ideal in Marine biology and fish farming. The combination of refrigeration system and air jacket heating system provides optimum temperature and humidity.
  • Refrigeration unit with connection to a compact table top refrigeration thermostat
  • Microprocessor control
  • Automatic start-up and automatic auto-zero adjustment of CO2 measuring cell
  • Independent electronic overtemperature protection with separate sensor
  • Heated external door and glass internal door
  • One-piece metal shelves