Anti-ADA Polyclonal Antibody Pair
ABNOH00000100-AP21
: Abnova
Specifications
- Antigen name:Adenosine Deaminase
- Antigen symbol:ADA
- Clonality:Polyclonal
- Host:Multiple
- Reactivity:Human
- Gene ID:100
- Antigen synonyms:Adenosine Aminohydrolase
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human ADA.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-ADA (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-ADA (20 µg);
Type:
Antigen: ADA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human