Italy
Order Entry
Accessories for CORIO series baths and circulators
Accessories for CORIO series baths and circulators
Supplier:  Julabo GmbH
Certificates
undefined
Accessories for CORIO series baths and circulators
Supplier:  Julabo GmbH
Related Information
Product Information
  • Catalog #
    Supplier #
    For
    Pk
    Availability
    Price
  • Plastic test tube rack for 60 tubes with 17 mm Ø, height 100 mm
    Plastic test tube rack for 60 tubes with 17 mm Ø, height 100 mm
    461-0941
    9970300
    B13, B17, B19, B27, B33, BT19, BT27, PURA™
    1
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 63,60 €
    63,60 €
    1 * 1 pezzi
       63,60 €
     : 9970300
     : JULA9970300
    Specifications:
    Specifications
    Environmentally Preferable
  • Plastic test tube rack for 90 tubes with 12/13 mm Ø, height 75 mm
    Plastic test tube rack for 90 tubes with 12/13 mm Ø, height 75 mm
    461-0942
    9970301
    B13, B17, B19, B27, B33, BT19, BT27, PURA™
    1
    1 * 1 pezzi
    1 * 1 pezzi - 63,60 €
    63,60 €
    1 * 1 pezzi
       63,60 €
     : 9970301
     : JULA9970301
    Specifications
    Environmentally Preferable
  • Plastic test tube rack for 21 tubes with 30 mm Ø
    Plastic test tube rack for 21 tubes with 30 mm Ø
    461-0943
    9970303
    B13, B17, B19, B27, B33, BT19, BT27, PURA™
    1
    1 * 1 pezzi
    1 * 1 pezzi - 63,60 €
    63,60 €
    1 * 1 pezzi
       63,60 €
     : 9970303
     : JULA9970303
    Specifications
    Environmentally Preferable
  • Stainless steel test tube rack for 30 tubes with 17 mm Ø, height 100 mm
    Stainless steel test tube rack for 30 tubes with 17 mm Ø, height 100 mm
    461-0944
    9970320
    B5, BT5
    1
    1 * 1 pezzi
    1 * 1 pezzi - 105,00 €
    105,00 €
    1 * 1 pezzi
       105,00 €
     : 9970320
     : JULA9970320
    Specifications
    Environmentally Preferable
  • Stainless steel test tube rack for 42 tubes with 12/13 mm Ø, height 75 mm
    Stainless steel test tube rack for 42 tubes with 12/13 mm Ø, height 75 mm
    461-0945
    9970321
    B5, BT5
    1
    1 * 1 pezzi
    1 * 1 pezzi - 105,00 €
    105,00 €
    1 * 1 pezzi
       105,00 €
     : 9970321
     : JULA9970321
    Specifications
    Environmentally Preferable
  • Stainless steel test tube rack for 42 tubes with 10/11 mm Ø, height 40 mm
    Stainless steel test tube rack for 42 tubes with 10/11 mm Ø, height 40 mm
    461-0946
    9970322
    B5, BT5
    1
    1 * 1 pezzi
    1 * 1 pezzi - 105,00 €
    105,00 €
    1 * 1 pezzi
       105,00 €
     : 9970322
     : JULA9970322
    Specifications
    Environmentally Preferable
  • Stainless steel test tube rack for 10×50 ml falcon tubes
    Stainless steel test tube rack for 10×50 ml falcon tubes
    461-0947
    9970323
    B5, BT5
    1
    1 * 1 pezzi
    1 * 1 pezzi - 105,00 €
    105,00 €
    1 * 1 pezzi
       105,00 €
     : 9970323
     : JULA9970323
    Specifications
    Environmentally Preferable
  • Immersion-height adjustable platform
    Immersion-height adjustable platform
    461-0950
    9970503
    B13, B17
    1
    1 * 1 pezzi
    1 * 1 pezzi - 346,00 €
    346,00 €
    1 * 1 pezzi
       346,00 €
     : 9970503
     : JULA9970503
    Specifications
    Environmentally Preferable
  • Bath lid with built-in heat exchanger
    Bath lid with built-in heat exchanger
    461-0931
    9970240
    BC4, BC6
    1
    1 * 1 pezzi
    1 * 1 pezzi - 446,00 €
    446,00 €
    1 * 1 pezzi
       446,00 €
     : 9970240
     : JULA9970240
    Specifications
    Environmentally Preferable
  • Immersion-height adjustable platform
    Immersion-height adjustable platform
    461-0949
    9970502
    B19, B27
    1
    1 * 1 pezzi
    1 * 1 pezzi - 361,00 €
    361,00 €
    1 * 1 pezzi
       361,00 €
     : 9970502
     : JULA9970502
    Specifications
    Environmentally Preferable
  • Bath lid with built-in heat exchanger
    Bath lid with built-in heat exchanger
    461-0932
    9970242
    BC12
    1
    1 * 1 pezzi
    1 * 1 pezzi - 520,00 €
    520,00 €
    1 * 1 pezzi
       520,00 €
     : 9970242
     : JULA9970242
    Specifications
    Environmentally Preferable
  • Bath lid, flat
    Bath lid, flat
    461-0940
    9970296
    B5, BT5
    1
    1 * 1 pezzi
    1 * 1 pezzi - 136,00 €
    136,00 €
    1 * 1 pezzi
       136,00 €
     : 9970296
     : JULA9970296
    Specifications
    Environmentally Preferable
  • Bath lid, flat
    Bath lid, flat
    461-0936
    9970290
    B13, B17
    1
    1 * 1 pezzi
    1 * 1 pezzi - 190,00 €
    190,00 €
    1 * 1 pezzi
       190,00 €
     : 9970290
     : JULA9970290
    Specifications
    Environmentally Preferable
  • Bath lid, flat
    Bath lid, flat
    461-0937
    9970291
    B19, B27, BT19, BT27
    1
    1 * 1 pezzi
    1 * 1 pezzi - 215,00 €
    215,00 €
    1 * 1 pezzi
       215,00 €
     : 9970291
     : JULA9970291
    Specifications
    Environmentally Preferable
  • Bath lid, flat
    Bath lid, flat
    461-0938
    9970292
    B33
    1
    1 * 1 pezzi
    1 * 1 pezzi - 287,00 €
    287,00 €
    1 * 1 pezzi
       287,00 €
     : 9970292
     : JULA9970292
    Specifications
    Environmentally Preferable
  • Bath lid, flat
    Bath lid, flat
    461-0939
    9970293
    B39
    1
    1 * 1 pezzi
    1 * 1 pezzi - 244,00 €
    244,00 €
    1 * 1 pezzi
       244,00 €
     : 9970293
     : JULA9970293
    Specifications
    Environmentally Preferable
  • Gabled stainless steel lift-up lid
    Gabled stainless steel lift-up lid
    461-0933
    9970253
    B13, B17
    1
    1 * 1 pezzi
    1 * 1 pezzi - 435,00 €
    435,00 €
    1 * 1 pezzi
       435,00 €
     : 9970253
     : JULA9970253
    Specifications
    Environmentally Preferable
  • Gabled stainless steel lift-up lid
    Gabled stainless steel lift-up lid
    461-0934
    9970254
    B19, B27, BT19
    1
    1 * 1 pezzi
    1 * 1 pezzi - 469,00 €
    469,00 €
    1 * 1 pezzi
       469,00 €
     : 9970254
     : JULA9970254
    Specifications
    Environmentally Preferable
  • Gabled stainless steel lift-up lid
    Gabled stainless steel lift-up lid
    461-0935
    9970257
    B33
    1
    1 * 1 pezzi
    1 * 1 pezzi - 477,00 €
    477,00 €
    1 * 1 pezzi
       477,00 €
     : 9970257
     : JULA9970257
    Specifications
    Environmentally Preferable
  • Stainless steel bridge including assembly frame
    Stainless steel bridge including assembly frame
    461-0930
    9970229
    C-B5, C-BT5
    1
    1 * 1 pezzi
    1 * 1 pezzi - 316,00 €
    316,00 €
    1 * 1 pezzi
       316,00 €
     : 9970229
     : JULA9970229
    Specifications
    Environmentally Preferable
  • Stainless steel bridge including assembly frame
    Stainless steel bridge including assembly frame
    461-0929
    9970228
    C-B13, C-B17, C-B19, C-B27, C-BT19, C-BT27
    1
    1 * 1 pezzi
    1 * 1 pezzi - 316,00 €
    316,00 €
    1 * 1 pezzi
       316,00 €
     : 9970228
     : JULA9970228
    Specifications
    Environmentally Preferable
  • Stand attachment for laboratory stands, with rod, 200×12 mm Ø
    Stand attachment for laboratory stands, with rod, 200×12 mm Ø
    461-0928
    9970022
    C, CD
    1
    1 * 1 pezzi
    1 * 1 pezzi - 63,60 €
    63,60 €
    1 * 1 pezzi
       63,60 €
     : 9970022
     : JULA9970022
    Specifications
    Environmentally Preferable
  • Bath attachment clamp, for bath wall thickeness < 30 mm
    Bath attachment clamp, for bath wall thickeness < 30 mm
    461-0948
    9970420
    C, CD
    1
    1 * 1 pezzi
    1 * 1 pezzi - 42,40 €
    42,40 €
    1 * 1 pezzi
       42,40 €
     : 9970420
     : JULA9970420
    Specifications
    Environmentally Preferable
  • USB cable, 2 m Type A-B
    USB cable, 2 m Type A-B
    461-0920
    9900110
    CORIO™ CD
    1
    1 * 1 pezzi
    1 * 1 pezzi - 24,40 €
    24,40 €
    1 * 1 pezzi
       24,40 €
     : 9900110
     : JULA9900110