Italy
Order Entry
Anti-FFAR1 Mouse Monoclonal Antibody [clone: G16]
Anti-FFAR1 Mouse Monoclonal Antibody [clone: G16]
  ABNOMAB1714
 :  Abnova
Anti-FFAR1 Mouse Monoclonal Antibody [clone: G16]
  ABNOMAB1714
 :  Abnova
 : MAB1714
 : ABNOMAB1714
 
1 040,00 €
   
  1 * 25 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    Free fatty acid receptor 1
  • Antigen symbol:
    FFAR1
  • Clonality:
    Monoclonal
  • Clone:
    G16
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    233081
  • Antigen synonyms:
    Gpr40
  • Storage buffer:
    PBS (0,1% proclin, 2.0% Block Ace)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to mouse Ffar1.
  • Size:
    25 μg
  • Pk:
    25 µg

 

 

Mouse monoclonal antibody raised against synthetic peptide of Ffar1.

Recommended Dilutions: Immunocytochemistry: 1 ?g/ml; Flow Cytometry: 0.5 ?g/ml

Type: Primary
Antigen: FFAR1
Clonality: Monoclonal
Clone: G16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse