Italy
Order Entry
Anti-TRPV1 Rabbit Polyclonal Antibody
  ABNOPAB0698
 :  Abnova
Anti-TRPV1 Rabbit Polyclonal Antibody
  ABNOPAB0698
 :  Abnova
 
402,00 €
   
  1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    Transient receptor potential cation channel subfamily V member 1
  • Antigen symbol:
    TRPV1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Gene ID:
    7442
  • Antigen synonyms:
    TrpV1|VR1|TRPV1|Transient receptor potential cation channel subfamily V member 1|Capsaicin receptor
  • Amino acid number:
    608 to 621
  • Storage buffer:
    PBS
  • Sequence:
    PSESTSHRWRGPA
  • Storage temperature:
    Store at 4 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide (conjugated with KLH) corresponding to amino acids 608-621 human TRPV1.
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody raised against synthetic peptide of TRPV1.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2000

Type: Primary
Antigen: TRPV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat