Anti-ABCA1 Monoclonal Antibody [clone: AB1.G6]
ABNOMAB8291
: Abnova
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family A, member 1
- Antigen symbol:ABCA1
- Clonality:Monoclonal
- Clone:AB1.G6
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:ABC-1|ATP-binding cassette transporter 1|CERP|HDLDT1|TGD|ABC1|membrane-bound|ATP-binding cassette transporter A1|cholesterol efflux regulatory protein
- Amino acid number:1800 to 2260
- Storage buffer:PBS, pH 7,4 (0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 1800-2260 of ABCA1.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against synthetic peptide of ABCA1.
Type: Primary
Antigen: ABCA1
Clonality: Monoclonal
Clone: AB1.G6
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: