Stampa…

Anti-ELMOD1 Rabbit Polyclonal Antibody

Fornitore: Abgent
ABGEAP18029CEA 348 EUR
ABGEAP18029C
Anti-ELMOD1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: ELMOD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR