Stampa…
Anti-YF005 Rabbit Polyclonal Antibody
Fornitore: Abgent
ABGEAP19207AEA 348 EUR
ABGEAP19207A
Anti-YF005 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: YF005
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human