Stampa…
Anti-AIM1L Rabbit Polyclonal Antibody
Fornitore: Abgent
ABGEAP16345BEA 348 EUR
ABGEAP16345B
Anti-AIM1L Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: AIM1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human