Stampa…

Anti-BEST3 Rabbit Polyclonal Antibody

Fornitore: Abgent
ABGEAP19190AEA 348 EUR
ABGEAP19190A
Anti-BEST3 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: BEST3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR