Stampa…

Anti-CARF Rabbit Polyclonal Antibody

Fornitore: Abgent
ABGEAP18673AEA 348 EUR
ABGEAP18673A
Anti-CARF Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: CARF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR