Stampa…
Anti-ZFP82 Rabbit Polyclonal Antibody
Fornitore: Abgent
ABGEAP17864AEA 348 EUR
ABGEAP17864A
Anti-ZFP82 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: ZFP82
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human