Stampa…

Anti-CNTROB Rabbit Polyclonal Antibody

Fornitore: Abgent
ABGEAP16471CEA 348 EUR
ABGEAP16471C
Anti-CNTROB Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: CNTROB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR