Stampa…
Anti-MIER1 Rabbit Polyclonal Antibody
Fornitore: Abgent
ABGEAP18957CEA 348 EUR
ABGEAP18957C
Anti-MIER1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: MIER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human