Stampa…

Anti-NRG1 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBI143581-50EA 324 EUR
USBI143581-50 USBI143581
Anti-NRG1 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 0.5-2?g/ml

Type: Primary
Antigen: NRG1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR