Italy
Order Entry
Anti-FtsZ Rabbit Polyclonal Antibody
  AGRIAS07217
 :  Agrisera
undefined
Anti-FtsZ Rabbit Polyclonal Antibody
  AGRIAS07217
 :  Agrisera
 
437,00 €
   
  1 * 200 µl

 

  • Antibody type:
    Primary
  • Antigen symbol:
    FTSZ
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    A. thaliana
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    whole cyanobacterial (Anabaena PCC 7120) FtsZ protein Q3MC27 overexpressed in E.coli.
  • Tested applications:
    Immunolocalization
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Type: Primary
Antigen: FTSZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana