Italy
Order Entry
Anti-FRZB Rabbit Polyclonal Antibody
Anti-FRZB Rabbit Polyclonal Antibody
  BSBTPB9372
undefined
Anti-FRZB Rabbit Polyclonal Antibody
  BSBTPB9372
 : PB9372
 : BSBTPB9372
 
552,00 €
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    Frizzled-Related Protein
  • Antigen symbol:
    FRZB
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Gene ID:
    2487
  • Antigen synonyms:
    FRZB-PEN|hFIZ|SFRP3|FRP-3|OS1|FRE|FRITZ|FZRB|FRZB1|SRFP3|FRZB-1
  • UniProtKB:
    Q92765
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    E.coli-derived human FRZB recombinant protein (Position: E176-N325). Human FRZB shares 90% amino acid (aa) sequence identity with mouse FRZB.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 µg/vial
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Secreted frizzled-related protein 3(FRZB) detection. Tested with WB, ELISA in Human.

FRZB is also known as FRE or OS1. The protein encoded by this gene is a secreted protein that is involved in the regulation of bone development. Defects in this gene are a cause of female-specific osteoarthritis (OA) susceptibility. FRZB is a Wnt-binding protein especially important in embryonic development. It is a competitor for the cell-surface G-protein receptor Frizzled. FRZB is localized in the extracellular plasma membrane. Unlike frizzled, frzb lacks the 7 transmembrane domains normally found in G-protein-coupled receptors. It is still considered a homolog of frizzled because it contains a Cysteine Rich Domain (CRD), and because of its intracellular C-terminus which is crucial for signaling.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

WB: The detection limit for FRZB is approximately 0.1ng/lane under reducing conditions.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: GRIN2B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
, Human