Stampa…
Goat Anti-Rabbit IgG Polyclonal Antibody (DyLight® 633)
IMMRIR2168EA 101 EUR
IMMRIR2168
Goat Anti-Rabbit IgG Polyclonal Antibody (DyLight® 633)
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 633
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit