Anti-IgG Goat Polyclonal Antibody
IMMRIR1912
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:Yes
- Format:Purified IgG
- Form:Clear, colorless liquid, 0,2 µm filtered
- Antibody fragment:F(ab')2
- Storage buffer:10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:2…8 °C
- Concentration:>4,5 mg/ml (E 1% at 280 nm = 13,0)
- Shipping temperature:2…8 °C
- Immunogen:Mouse IgG
- Tested applications:Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
- Purification:Affinity purified using solid phaseMouse IgG
- Size:1,0 mg
- Pk:0,5 mg