Italy
Order Entry
Anti-IgA/IgG/IgM Goat Polyclonal Antibody
  IMMRIR1746
undefined
Anti-IgA/IgG/IgM Goat Polyclonal Antibody
  IMMRIR1746
Price
73,90 €
Log in for availability
  1 * 1 mg

 

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin A, G And M
  • Antigen symbol:
    IgG+IgA+IgM
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Clear, colorless liquid, 0,2 µm filtered
  • Storage buffer:
    10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2
  • Storage temperature:
    2…8 °C
  • Concentration:
    >4,5 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2…8 °C
  • Immunogen:
    Human IgG + IgA + IgM
  • Tested applications:
    Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
  • Purification:
    Affinity purified using solid phase Human IgA + IgG + IgM
  • Size:
    2,0 mg
  • Pk:
    1 mg

 

About this item