Stampa…

Anti-SOCS3 Rabbit polyclonal antibody

USBIS1040-03SEA 352 EUR
USBIS1040-03S
Anti-SOCS3 Rabbit polyclonal antibody
Recommended dilution: Western Blot: 0.5-2μg/ml; Immunochemistry: 2.5-20 μg/ml

Type: Primary
Antigen: SOCS3 (suppressor of cytokine signaling 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR