Stampa…

Anti-SOCS3 Mouse Monoclonal Antibody [clone: SO1]

Fornitore: United States Biological
USBIS1040-03REA 647 EUR
USBIS1040-03R
Anti-SOCS3 Mouse Monoclonal Antibody [clone: SO1]
Anticorpi
Recommended dilution: ELISA: 1:100-1:500; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: SOCS3 (suppressor of cytokine signaling 3)
Clonality: Monoclonal
Clone: SO1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR