Stampa…

Anti-SNURF Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIS1014-99EA 587 EUR
USBIS1014-99
Anti-SNURF Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: SNURF (SNRPN upstream reading frame)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR