Stampa…
Anti-ARHGAP33 Goat Polyclonal Antibody
Fornitore: United States Biological
USBIS1014-97T5EA 633 EUR
USBIS1014-97T5
Anti-ARHGAP33 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:32000; Immunochemistry: 1. 25 ?g/ml
Type: Primary
Antigen: ARHGAP33 (Rho GTPase activating protein 33)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Goat
Isotype:
Reactivity: Human