Stampa…

Anti-ARHGAP33 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIS1014-97T4EA 633 EUR
USBIS1014-97T4
Anti-ARHGAP33 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:8000; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: ARHGAP33 (Rho GTPase activating protein 33)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR